Assay Details
Gene Symbol: GADD45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299957.2 | XP_015155443.1 | 2 - 3 | 309 | 65 | |
GenBank mRNA | BX929568.2 | - | 2 - 3 | 308 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
