Assay Details
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001233782.5 | XP_001233783.1 | 1 - 2 | 202 | 68 | |
GenBank mRNA | BX929575.1 | - | 1 - 2 | 207 | 68 | |
BX932648.1 | - | 1 - 2 | 201 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
