Assay Details
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641052.4 | XP_003641100.1 | 5 - 6 | 442 | 112 | |
XM_015284995.2 | XP_015140481.1 | 5 - 6 | 2306 | 112 | ||
XM_015284998.2 | XP_015140484.1 | 4 - 5 | 2140 | 112 | ||
XM_025149115.1 | XP_025004883.1 | 5 - 6 | 2235 | 112 | ||
XM_025149116.1 | XP_025004884.1 | 4 - 5 | 2217 | 112 | ||
XM_025149118.1 | XP_025004886.1 | 5 - 6 | 513 | 112 | ||
GenBank mRNA | CR353756.1 | - | 5 - 6 | 407 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
