Assay Details
Gene Symbol: PRRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015278538.2 | XP_015134024.1 | 2 - 3 | 97 | 93 | |
GenBank mRNA | CR386888.1 | - | 2 - 3 | 83 | 93 | |
CR407157.1 | - | 2 - 3 | 79 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015278538.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
