Assay Details
Gene Symbol: FAM134C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234117.5 | XP_001234118.1 | 6 - 7 | 772 | 98 | |
GenBank mRNA | CR387665.1 | - | 3 - 4 | 1310 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
