Assay Details
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942343.3 | XP_004942400.2 | 1 - 2 | 136 | 80 | |
XM_015289064.2 | XP_015144550.2 | 1 - 2 | 136 | 80 | ||
XM_424063.6 | XP_424063.4 | 1 - 2 | 140 | 80 | ||
GenBank mRNA | CR387710.1 | - | 1 - 2 | 144 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
