Assay Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295293.2 | XP_015150779.1 | 3 - 4 | 5070 | 73 | |
GenBank mRNA | CR388525.1 | - | 3 - 4 | 388 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
