Assay Details
Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR389038.1 | - | 3 - 4 | 217 | 85 | |
CR389043.1 | - | 664 | 85 | |||
CR390830.1 | - | 3 - 4 | 298 | 85 | ||
CR407059.1 | - | 3 - 4 | 287 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
