Assay Details
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417855.6 | XP_417855.2 | 9 - 10 | 1184 | 104 | |
GenBank mRNA | CR389704.1 | - | 1 - 2 | 67 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
