Assay Details
Gene Symbol: NRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298695.2 | XP_015154181.1 | 4 | 8088 | 85 | |
XM_015298704.2 | XP_015154190.1 | 4 | 8196 | 85 | ||
XM_015298712.1 | XP_015154198.1 | 4 | 8006 | 85 | ||
XM_015298715.2 | XP_015154201.1 | 3 | 7923 | 85 | ||
XM_015298718.2 | XP_015154204.1 | 5 | 8214 | 85 | ||
XM_015298725.2 | XP_015154211.1 | 6 | 8311 | 85 | ||
XM_015298731.2 | XP_015154217.1 | 3 | 7985 | 85 | ||
XM_015298735.2 | XP_015154221.1 | 3 | 7996 | 85 | ||
XM_015298743.2 | XP_015154229.1 | 3 | 10560 | 85 | ||
XM_025144775.1 | XP_025000543.1 | 5 | 10799 | 85 | ||
XM_025144778.1 | XP_025000546.1 | 5 | 8168 | 85 | ||
XM_025144781.1 | XP_025000549.1 | 7 | 11022 | 85 | ||
XM_025144787.1 | XP_025000555.1 | 5 | 10783 | 85 | ||
GenBank mRNA | CR390637.1 | - | 1 | 926 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
