Assay Details
Gene Symbol: ARPC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279852.2 | XP_015135338.1 | 3 - 4 | 419 | 171 | |
GenBank mRNA | BX929285.2 | - | 3 - 4 | 401 | 171 | |
BX930304.1 | - | 3 - 4 | 403 | 171 | ||
BX933204.1 | - | 3 - 4 | 406 | 171 | ||
CR338728.2 | - | 3 - 4 | 735 | 171 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
