Assay Details
Gene Symbol: GAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004940820.3 | XP_004940877.1 | 10 | 5927 | 93 | |
XM_004940821.2 | XP_004940878.1 | 10 | 5923 | 93 | ||
XM_004940823.2 | XP_004940880.1 | 9 | 5815 | 93 | ||
XM_015278621.2 | XP_015134107.1 | 11 | 5984 | 93 | ||
GenBank mRNA | CR391508.1 | - | 1 | 1316 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
