Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX930182.2 | - | 3 - 4 | 337 | 123 | |
BX930766.1 | - | 3 - 4 | 329 | 123 | ||
BX932061.1 | - | 3 - 4 | 367 | 123 | ||
CR387027.1 | - | 2 - 3 | 264 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
