Assay Details
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275690.2 | XP_015131176.1 | 2 - 3 | 557 | 99 | |
GenBank mRNA | BX930279.2 | - | 1 - 2 | 459 | 99 | |
BX932214.2 | - | 2 - 3 | 240 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
