Assay Details
Gene Symbol: YPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415068.6 | XP_415068.1 | 1 - 2 | 297 | 100 | |
GenBank mRNA | CR523261.1 | - | 1 - 2 | 218 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_415068.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
