Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145677.1 | XP_025001445.1 | 4 - 5 | 673 | 59 | |
XM_025145678.1 | XP_025001446.1 | 4 - 5 | 666 | 59 | ||
XM_025145679.1 | XP_025001447.1 | 4 - 5 | 666 | 59 | ||
XM_025145680.1 | XP_025001448.1 | 4 - 5 | 668 | 59 | ||
GenBank mRNA | CR523931.1 | - | 2 - 3 | 228 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
