Assay Details
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_423779.6 | XP_423779.5 | 9 - 10 | 1346 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_423779.6 | XP_423779.5 | 9 - 10 | 1346 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |