Assay Details
Gene Symbol: GPR183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231943.4 | XP_001231944.1 | 1 - 2 | 1431 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001231943.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
