Assay Details
Gene Symbol: SRD5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015285539.2 | XP_015141025.1 | 2 - 3 | 403 | 96 | |
XM_420703.6 | XP_420703.2 | 2 - 3 | 389 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
