Assay Details
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945280.3 | XP_004945337.1 | 3 - 4 | 2529 | 68 | |
XM_004945281.3 | XP_004945338.1 | 3 - 4 | 2529 | 68 | ||
XM_004945282.3 | XP_004945339.1 | 3 - 4 | 2529 | 68 | ||
XM_004945283.3 | XP_004945340.1 | 3 - 4 | 2529 | 68 | ||
XM_015294525.2 | XP_015150011.1 | 3 - 4 | 2529 | 68 | ||
XM_025155170.1 | XP_025010938.1 | 2 - 3 | 2420 | 68 | ||
XM_414889.5 | XP_414889.1 | 3 - 4 | 2529 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
