Assay Details
Gene Symbol: TTLL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293350.2 | XP_015148836.1 | 2 - 3 | 131 | 99 | |
XM_015293351.2 | XP_015148837.1 | 2 - 3 | 129 | 99 | ||
XM_015293352.2 | XP_015148838.1 | 2 - 3 | 139 | 99 | ||
XM_025154714.1 | XP_025010482.1 | 11 - 12 | 1918 | 99 | ||
XM_025154717.1 | XP_025010485.1 | 2 - 3 | 126 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
