Assay Details
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641434.4 | XP_003641482.2 | 1 - 2 | 274 | 158 | |
XM_004936329.3 | XP_004936386.1 | 1 - 2 | 3483 | 158 | ||
XM_004936334.3 | XP_004936391.1 | 1 - 2 | 3483 | 158 | ||
XM_025151254.1 | XP_025007022.1 | 1 - 2 | 3483 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
