Assay Details
Gene Symbol: TSTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938354.3 | XP_004938411.1 | 1 - 2 | 92 | 120 | |
XM_416681.6 | XP_416681.3 | 1 - 2 | 92 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
