Assay Details
Gene Symbol: SLC2A11L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945642.3 | XP_004945699.1 | 9 - 10 | 1189 | 110 | |
XM_004945643.3 | XP_004945700.1 | 8 - 9 | 1049 | 110 | ||
XM_015275482.1 | XP_015130968.1 | 9 - 10 | 1135 | 110 | ||
XM_015275484.2 | XP_015130970.1 | 8 - 9 | 1105 | 110 | ||
XM_015275485.2 | XP_015130971.1 | 7 - 8 | 1003 | 110 | ||
XM_025155583.1 | XP_025011351.1 | 8 - 9 | 2406 | 110 | ||
XM_025155584.1 | XP_025011352.1 | 8 - 9 | 1020 | 110 | ||
XM_415207.6 | XP_415207.2 | 8 - 9 | 1513 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
