Assay Details
Gene Symbol: HUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275914.2 | XP_015131400.1 | 3 - 4 | 673 | 91 | |
XM_015275916.2 | XP_015131402.1 | 3 - 4 | 693 | 91 | ||
XM_025147602.1 | XP_025003370.1 | 3 - 4 | 674 | 91 | ||
XM_025147603.1 | XP_025003371.1 | 3 - 4 | 674 | 91 | ||
XM_025147604.1 | XP_025003372.1 | 2 - 3 | 197 | 91 | ||
XM_426029.6 | XP_426029.3 | 2 - 3 | 240 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
