Assay Details
Gene Symbol: C18H17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946179.3 | XP_004946236.2 | 6 - 7 | 923 | 131 | |
XM_004946180.3 | XP_004946237.2 | 5 - 6 | 853 | 131 | ||
XM_415664.6 | XP_415664.3 | 6 - 7 | 924 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
