Assay Details
Gene Symbol: C18H17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946179.3 | XP_004946236.2 | 8 - 9 | 1124 | 97 | |
XM_004946180.3 | XP_004946237.2 | 7 - 8 | 1054 | 97 | ||
XM_415664.6 | XP_415664.3 | 8 - 9 | 1125 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
