Assay Details
Gene Symbol: AKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287881.2 | XP_015143367.1 | 2 | 5691 | 104 | |
XM_025151017.1 | XP_025006785.1 | 3 | 5865 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
