Assay Details
Gene Symbol: ADGRG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015292332.2 | XP_015147818.2 | 5 - 6 | 847 | 100 | |
XM_015292333.2 | XP_015147819.2 | 5 - 6 | 843 | 100 | ||
XM_015292334.2 | XP_015147820.1 | 6 - 7 | 1037 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
