Assay Details
Gene Symbol: KCNJ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004934546.3 | XP_004934603.1 | 2 - 3 | 380 | 83 | |
XM_015301389.2 | XP_015156875.1 | 3 - 4 | 209 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004934546.3, XM_015301389.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
