Assay Details
Gene Symbol: USP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294380.2 | XP_015149866.1 | 16 - 17 | 6506 | 75 | |
XM_015294382.2 | XP_015149868.2 | 12 - 13 | 1372 | 75 | ||
XM_015294383.2 | XP_015149869.1 | 17 - 18 | 6551 | 75 | ||
XM_025155364.1 | XP_025011132.1 | 17 - 18 | 7577 | 75 | ||
XM_025155365.1 | XP_025011133.1 | 14 - 15 | 1654 | 75 | ||
XM_025155366.1 | XP_025011134.1 | 15 - 16 | 6362 | 75 | ||
XM_414805.6 | XP_414805.3 | 13 - 14 | 1486 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
