Assay Details
Gene Symbol: USP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294380.2 | XP_015149866.1 | 6 - 7 | 5276 | 132 | |
XM_015294382.2 | XP_015149868.2 | 2 - 3 | 241 | 132 | ||
XM_015294383.2 | XP_015149869.1 | 6 - 7 | 5276 | 132 | ||
XM_025155364.1 | XP_025011132.1 | 6 - 7 | 6302 | 132 | ||
XM_025155365.1 | XP_025011133.1 | 3 - 4 | 379 | 132 | ||
XM_025155366.1 | XP_025011134.1 | 6 - 7 | 5276 | 132 | ||
XM_414805.6 | XP_414805.3 | 2 - 3 | 211 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
