Assay Details
Gene Symbol: USP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294380.2 | XP_015149866.1 | 12 - 13 | 6065 | 85 | |
XM_015294382.2 | XP_015149868.2 | 8 - 9 | 931 | 85 | ||
XM_015294383.2 | XP_015149869.1 | 13 - 14 | 6110 | 85 | ||
XM_025155364.1 | XP_025011132.1 | 13 - 14 | 7136 | 85 | ||
XM_025155365.1 | XP_025011133.1 | 10 - 11 | 1213 | 85 | ||
XM_025155366.1 | XP_025011134.1 | 11 - 12 | 5921 | 85 | ||
XM_414805.6 | XP_414805.3 | 9 - 10 | 1045 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
