Assay Details
Gene Symbol: CPSF4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234821.5 | XP_001234822.3 | 3 - 4 | 357 | 69 | |
XM_015279937.2 | XP_015135423.1 | 2 - 3 | 261 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
