Assay Details
Gene Symbol: VGLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025144693.1 | XP_025000461.1 | 3 - 4 | 2378 | 126 | |
XM_416671.6 | XP_416671.3 | 3 - 4 | 975 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
