Assay Details
Gene Symbol: TNFAIP8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025153895.1 | XP_025009663.1 | 3 | 2674 | 82 | |
XM_425071.6 | XP_425071.3 | 2 | 2663 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
