Assay Details
Gene Symbol: RNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935386.3 | XP_004935443.1 | 3 - 4 | 1113 | 90 | |
XM_015283790.2 | XP_015139276.1 | 3 - 4 | 1571 | 90 | ||
XM_015283791.2 | XP_015139277.1 | 3 - 4 | 1573 | 90 | ||
XM_025148867.1 | XP_025004635.1 | 4 - 5 | 1039 | 90 | ||
XM_419487.4 | XP_419487.2 | 3 - 4 | 1032 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
