Assay Details
Gene Symbol: FAM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284934.2 | XP_015140420.1 | 1 - 2 | 209 | 96 | |
XM_015284935.2 | XP_015140421.1 | 1 - 2 | 7368 | 96 | ||
XM_025148866.1 | XP_025004634.1 | 1 - 2 | 7368 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015284934.2, XM_015284935.2, XM_025148866.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
