Assay Details
Gene Symbol: FAM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284934.2 | XP_015140420.1 | 2 - 3 | 520 | 145 | |
XM_015284935.2 | XP_015140421.1 | 2 - 3 | 7679 | 145 | ||
XM_025148866.1 | XP_025004634.1 | 2 - 3 | 7679 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
