Assay Details
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293879.2 | XP_015149365.1 | 5 - 6 | 1186 | 75 | |
GenBank mRNA | BX931053.2 | - | 4 - 5 | 391 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
