Assay Details
Gene Symbol: LOC107055017 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143030.1 | XP_024998798.1 | 2 | 2369 | 70 |
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297723.2 | XP_015153209.1 | 5 | 2784 | 70 | |
XM_015297724.2 | XP_015153210.1 | 5 | 2620 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
