Assay Details
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642446.4 | XP_003642494.3 | 5 - 6 | 3436 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
