Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287788.2 | XP_015143274.1 | 10 | 2609 | 128 | |
XM_015287789.2 | XP_015143275.1 | 10 | 2542 | 128 | ||
XM_015287790.2 | XP_015143276.1 | 9 | 2153 | 128 | ||
XM_015287791.2 | XP_015143277.1 | 9 | 2558 | 128 | ||
XM_015287792.2 | XP_015143278.1 | 9 | 2517 | 128 | ||
XM_015287793.2 | XP_015143279.1 | 8 | 2057 | 128 | ||
XM_015287794.2 | XP_015143280.1 | 8 | 2476 | 128 | ||
XM_025151157.1 | XP_025006925.1 | 6 | 2318 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
