Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938375.3 | XP_004938432.1 | 8 - 9 | 1363 | 94 | |
XM_004938376.3 | XP_004938433.1 | 5 - 6 | 824 | 94 | ||
XM_015299566.2 | XP_015155052.1 | 7 - 8 | 1283 | 94 | ||
XM_015299571.2 | XP_015155057.1 | 6 - 7 | 599 | 94 | ||
XM_025144927.1 | XP_025000695.1 | 7 - 8 | 1455 | 94 | ||
XM_025144929.1 | XP_025000697.1 | 6 - 7 | 1294 | 94 | ||
XM_025144931.1 | XP_025000699.1 | 7 - 8 | 1029 | 94 | ||
XM_025144936.1 | XP_025000704.1 | 4 - 5 | 351 | 94 | ||
XM_025144938.1 | XP_025000706.1 | 5 - 6 | 1330 | 94 | ||
XM_025144941.1 | XP_025000709.1 | 3 - 4 | 287 | 94 | ||
GenBank mRNA | BX932098.2 | - | 5 - 6 | 802 | 94 | |
BX933111.2 | - | 2 - 3 | 186 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
