Assay Details
Gene Symbol: UMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281905.2 | XP_015137391.1 | 4 - 5 | 408 | 88 | |
GenBank mRNA | BX932167.1 | - | 4 - 5 | 308 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
