Assay Details
Gene Symbol: GDF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145635.1 | XP_025001403.1 | 2 - 3 | 1009 | 103 | |
GenBank mRNA | LC274960.1 | - | 2 - 3 | 613 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
