Assay Details
Gene Symbol: MECP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | Y14166.1 | - | 90 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the Y14166.1 transcripts.
Set Membership: |
![]() ![]() |
