Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945570.3 | XP_004945627.3 | 2 - 3 | 287 | 63 | |
XM_015275582.2 | XP_015131068.1 | 1 - 2 | 556 | 63 | ||
XM_015275583.2 | XP_015131069.1 | 2 - 3 | 311 | 63 | ||
XM_415152.6 | XP_415152.4 | 2 - 3 | 213 | 63 | ||
GenBank mRNA | BX932562.1 | - | 1 - 2 | 551 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
