Assay Details
Gene Symbol: MCRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422971.6 | XP_422971.3 | 2 - 3 | 212 | 106 | |
GenBank mRNA | BX933195.2 | - | 2 - 3 | 194 | 106 | |
CR385394.1 | - | 2 - 3 | 313 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
