Assay Details
Gene Symbol: MCRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422971.6 | XP_422971.3 | 3 - 4 | 100 | ||
GenBank mRNA | BX933195.2 | - | 3 - 4 | 325 | 100 |
Gene Symbol: WFIKKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025155291.1 | XP_025011059.1 | 3 - 4 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
